Product Details

SNP ID
rs35822095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36339214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATGTCTACAGATCTTCCTTTTA[C/T]GTGAATTCACTAATACTAATGACAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00665 PubMed Links

Gene Details

Gene
LINC00665
Gene Name
long intergenic non-protein coding RNA 665
There are no transcripts associated with this gene.

Gene
ZFP14
Gene Name
ZFP14 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297619.1 2735 UTR 3 NP_001284548.1
NM_020917.2 2735 UTR 3 NP_065968.1
XM_017027038.1 2735 UTR 3 XP_016882527.1

View Full Product Details