Product Details
- SNP ID
-
rs16987029
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56177201 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATCCGCACCTGCCCACCGGGTAAC[A/G]CCTCCCTAAGTTCCTCGGAAACAAC
- Phenotype
-
MIM: 611178
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GALP
PubMed Links
Gene Details
- Gene
- GALP
- Gene Name
- galanin like peptide
View Full Product Details