Product Details

SNP ID
rs17130638
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:88855880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTTGGCCAGGAGGCTGGTCACC[A/G]CACCTAGTTTATGAATTTCTTTACA
Phenotype
MIM: 189963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF2B PubMed Links

Gene Details

Gene
GTF2B
Gene Name
general transcription factor IIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001514.5 Intron NP_001505.1
XM_011541299.1 Intron XP_011539601.1

View Full Product Details