Product Details

SNP ID
rs17130636
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:88854860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTGATTCAAATCAGTGAGGTTTT[A/G]GTGATTTTCACTGTGAATGCCTGCC
Phenotype
MIM: 189963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF2B PubMed Links

Gene Details

Gene
GTF2B
Gene Name
general transcription factor IIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001514.5 Intron NP_001505.1
XM_011541299.1 Intron XP_011539601.1

View Full Product Details