Product Details

SNP ID
rs17130577
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:88695248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCTTTATTAGCCTAACTCTAACT[G/T]GCAAAAATTAATCAAATATGATTAA
Phenotype
MIM: 602549
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PKN2 PubMed Links

Gene Details

Gene
PKN2
Gene Name
protein kinase N2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320707.1 Intron NP_001307636.1
NM_001320708.1 Intron NP_001307637.1
NM_001320709.1 Intron NP_001307638.1
NM_006256.3 Intron NP_006247.1
XM_011541772.2 Intron XP_011540074.1
XM_017001782.1 Intron XP_016857271.1
XM_017001783.1 Intron XP_016857272.1

View Full Product Details