Product Details

SNP ID
rs17856235
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61804753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGTAGAGAGGCAGCAAGGCTG[A/G]GGGCCCAACTGGGGAGGAAACCGAG
Phenotype
MIM: 606148 MIM: 600393
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013402.4 787 Missense Mutation CCA,TCA P329S NP_037534.3
XM_011545022.2 787 Missense Mutation CCA,TCA P258S XP_011543324.1
Gene
FEN1
Gene Name
flap structure-specific endonuclease 1
There are no transcripts associated with this gene.

View Full Product Details