Product Details
- SNP ID
-
rs17128262
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:55153050 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTTACTCGTTACTTGTTAATCCA[A/G]GTACCTGCTAAGAAACTTGACTTCA
- Phenotype
-
MIM: 153619
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DLGAP5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148827496] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DLGAP5
- Gene Name
- DLG associated protein 5
- Gene
- LGALS3
- Gene Name
- lectin, galactoside binding soluble 3
There are no transcripts associated with this gene.
View Full Product Details