Product Details

SNP ID
rs17128829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55790489 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAGCACAAAACAGACTAAAGCCA[A/G]TTTGCACACTTGGTTCCTTGGCTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00520 PubMed Links

Gene Details

Gene
LINC00520
Gene Name
long intergenic non-protein coding RNA 520
There are no transcripts associated with this gene.

View Full Product Details