Product Details

SNP ID
rs17129144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73658731 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTAAAGAATTATTAAACAGTTT[A/G]CTAAATTCTAGCAATTGTCTTGAAA
Phenotype
MIM: 610062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAL1 PubMed Links

Gene Details

Gene
DNAL1
Gene Name
dynein axonemal light chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201366.1 Intron NP_001188295.1
NM_031427.3 Intron NP_113615.2
XM_017021679.1 Intron XP_016877168.1

View Full Product Details