Product Details

SNP ID
rs17849833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177455331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGTAGCCGCCTGGAGGCGCCGGC[G/T]CACGCGCACTCGCATCCGCACGTGC
Phenotype
MIM: 126660
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DBN1 PubMed Links

Gene Details

Gene
DBN1
Gene Name
drebrin 1
There are no transcripts associated with this gene.

Gene
PRR7
Gene Name
proline rich 7 (synaptic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174101.1 1462 Silent Mutation GCG,GCT A88A NP_001167572.1
NM_001174102.1 1462 Silent Mutation GCG,GCT A88A NP_001167573.1
NM_030567.4 1462 Silent Mutation GCG,GCT A88A NP_085044.2
XM_011534662.1 1462 Silent Mutation GCG,GCT A88A XP_011532964.1
XM_011534663.2 1462 Silent Mutation GCG,GCT A88A XP_011532965.1
XM_017009896.1 1462 Silent Mutation GCG,GCT A88A XP_016865385.1
XM_017009897.1 1462 Silent Mutation GCG,GCT A88A XP_016865386.1
Gene
PRR7-AS1
Gene Name
PRR7 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details