Product Details

SNP ID
rs16826463
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:39679814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCCCAGTTCCTGGCCTGGCAT[C/T]GTGTGTTCACACTGCTGGGCTGTCT
Phenotype
MIM: 610525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPCAL4 PubMed Links

Gene Details

Gene
HPCAL4
Gene Name
hippocalcin like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282396.1 3711 UTR 3 NP_001269325.1
NM_001282397.1 3711 UTR 3 NP_001269326.1
NM_016257.3 3711 UTR 3 NP_057341.1
XM_017001447.1 3711 UTR 3 XP_016856936.1
Gene
NT5C1A
Gene Name
5'-nucleotidase, cytosolic IA
There are no transcripts associated with this gene.

View Full Product Details