Product Details

SNP ID
rs16878911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87283999 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTCTGTCTAGAGCTCATAACTGA[A/C]AGCATATTGACAATGTAAAAAAATT
Phenotype
MIM: 611921
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GJB7 PubMed Links

Gene Details

Gene
GJB7
Gene Name
gap junction protein beta 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198568.2 1260 UTR 3 NP_940970.1
XM_005248716.2 1260 UTR 3 XP_005248773.1

View Full Product Details