Product Details

SNP ID
rs17230826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38742507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGATCATCATGGCAGCTATGTT[G/T]GTCCCATGTCCCTGGACTCTGCTGG
Phenotype
MIM: 611723
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00114 PubMed Links

Gene Details

Gene
LINC00114
Gene Name
long intergenic non-protein coding RNA 114
There are no transcripts associated with this gene.

View Full Product Details