Product Details
- SNP ID
-
rs16832811
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:161316699 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGACACCTCCATTTCTCATAAGGG[A/C]TTTTTTTAGGAGGATAAATGAGGAA
- Phenotype
-
MIM: 159440
MIM: 602413
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MPZ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116339979] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MPZ
- Gene Name
- myelin protein zero
There are no transcripts associated with this gene.
- Gene
- SDHC
- Gene Name
- succinate dehydrogenase complex subunit C
View Full Product Details