Product Details

SNP ID
rs17250163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19063884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATACCCTCGGATCTCCTCCAGC[C/G]CTAATACTCAGGTGACTACTTGGTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TTTY14 PubMed Links

Gene Details

Gene
TTTY14
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details