Product Details

SNP ID
rs17027687
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:111452693 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTCAGCCTAAGCATACCATTTA[C/G]TGCTATGATTTAAATCAGTGCTTCT
Phenotype
MIM: 603270 MIM: 611734
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP5F1 PubMed Links

Gene Details

Gene
ATP5F1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001688.4 Intron NP_001679.2
Gene
WDR77
Gene Name
WD repeat domain 77
There are no transcripts associated with this gene.

View Full Product Details