Product Details

SNP ID
rs17642803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99668558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCTTTTAGTCTTGTCATTTGC[A/C]TAAGAAGAGTAGAGCATCTAGGAAT
Phenotype
MIM: 616753 MIM: 609767
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ENTPD7 PubMed Links

Gene Details

Gene
ENTPD7
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020354.3 Intron NP_065087.1
Gene
SLC25A28
Gene Name
solute carrier family 25 member 28
There are no transcripts associated with this gene.

View Full Product Details