Product Details

SNP ID
rs17480775
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89582214 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACCCAGACTGAAGCTGCCTCAC[A/G]ATAAAAAGCCATGCTCTTACTTCAT
Phenotype
MIM: 606160
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PANK1 PubMed Links

Gene Details

Gene
PANK1
Gene Name
pantothenate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138316.3 4259 UTR 3 NP_612189.2
NM_148977.2 4259 UTR 3 NP_683878.1
NM_148978.2 4259 UTR 3 NP_683879.1
XM_017016333.1 4259 Intron XP_016871822.1
XM_017016334.1 4259 Intron XP_016871823.1
XM_017016335.1 4259 Intron XP_016871824.1
XM_017016336.1 4259 Intron XP_016871825.1
XM_017016337.1 4259 Intron XP_016871826.1
XM_017016338.1 4259 Intron XP_016871827.1

View Full Product Details