Product Details

SNP ID
rs34986991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:171420420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCCCGCAGGTGTTTACACTTC[A/C]TGCTGCTGTGCTTCCAGGTACAGGT
Phenotype
MIM: 603726 MIM: 164040
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FGF18 PubMed Links

Gene Details

Gene
FGF18
Gene Name
fibroblast growth factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003862.2 583 Missense Mutation ATG,CTG M16L NP_003853.1
Gene
NPM1
Gene Name
nucleophosmin
There are no transcripts associated with this gene.

View Full Product Details