Product Details

SNP ID
rs17055392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:55466672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAATTTTTTATTAATTTTCTTGT[A/G]TTGGGAAGATCTTGAATACGCTCCA
Phenotype
MIM: 164975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT5A PubMed Links

Gene Details

Gene
WNT5A
Gene Name
Wnt family member 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256105.1 4772 UTR 3 NP_001243034.1
NM_003392.4 4772 UTR 3 NP_003383.2
XM_006713324.1 4772 UTR 3 XP_006713387.1
XM_011534085.2 4772 UTR 3 XP_011532387.1
XM_011534086.2 4772 UTR 3 XP_011532388.1
XM_011534087.2 4772 UTR 3 XP_011532389.1
XM_011534088.2 4772 UTR 3 XP_011532390.1
XM_011534089.1 4772 UTR 3 XP_011532391.1
XM_017007127.1 4772 UTR 3 XP_016862616.1
XM_017007128.1 4772 UTR 3 XP_016862617.1

View Full Product Details