Product Details
- SNP ID
-
rs17122060
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118308327 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAGGGAATTGGAAGATCCATTGTT[C/T]CCTAAGAGTTGAGGGAAGAGTCCCA
- Phenotype
-
MIM: 186830
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD3E
PubMed Links
Gene Details
- Gene
- CD3E
- Gene Name
- CD3e molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000733.3 |
|
Intron |
|
|
NP_000724.1 |
View Full Product Details