Product Details

SNP ID
rs17305269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54075076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTGCTGTTGGCAGGGACCACCG[A/G]GCTGGGCCAGGCACTGAGGGACGGC
Phenotype
MIM: 616802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TARM1 PubMed Links

Gene Details

Gene
TARM1
Gene Name
T cell-interacting, activating receptor on myeloid cells 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135686.2 134 Missense Mutation CCG,TCG P37S NP_001129158.2
XM_005278278.3 134 Intron XP_005278335.1

View Full Product Details