Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286577.1 | Intron | NP_001273506.1 | |||
NM_015531.5 | Intron | NP_056346.3 | |||
XM_011544904.1 | Intron | XP_011543206.1 | |||
XM_011544905.1 | Intron | XP_011543207.1 | |||
XM_017017510.1 | Intron | XP_016872999.1 | |||
XM_017017511.1 | Intron | XP_016873000.1 | |||
XM_017017512.1 | Intron | XP_016873001.1 | |||
XM_017017513.1 | Intron | XP_016873002.1 | |||
XM_017017514.1 | Intron | XP_016873003.1 | |||
XM_017017515.1 | Intron | XP_016873004.1 | |||
XM_017017516.1 | Intron | XP_016873005.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271593.1 | Intron | NP_001258522.1 | |||
NM_016147.2 | Intron | NP_057231.1 | |||
XM_017017913.1 | Intron | XP_016873402.1 |