Product Details

SNP ID
rs17149941
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87835529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGTTTGACTGAAGTGAACATT[C/T]GCTACTATCAGTAAAAAACTCCCTC
Phenotype
MIM: 610821
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RUNDC3B PubMed Links

Gene Details

Gene
RUNDC3B
Gene Name
RUN domain containing 3B
There are no transcripts associated with this gene.

Gene
SLC25A40
Gene Name
solute carrier family 25 member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018843.3 2112 UTR 3 NP_061331.2

View Full Product Details