Product Details

SNP ID
rs16962079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49127833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGATCAAAAAAGCACTTCTGCCA[C/T]AACTCCAATAATTTTCAGGACACAT
Phenotype
MIM: 604508
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPS2 PubMed Links

Gene Details

Gene
COPS2
Gene Name
COP9 signalosome subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143887.1 1549 UTR 3 NP_001137359.1
NM_004236.3 1549 UTR 3 NP_004227.1

View Full Product Details