Product Details

SNP ID
rs17840403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64898919 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAATGGCATTTGTTCCCCATAAG[C/T]TTTCTACAATTATTTGCTTTGCTGA
Phenotype
MIM: 604756
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC441239 PubMed Links

Gene Details

Gene
LOC441239
Gene Name
uncharacterized LOC441239
There are no transcripts associated with this gene.

Gene
ZNF273
Gene Name
zinc finger protein 273
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021148.2 Intron NP_066971.2
XM_011515746.2 Intron XP_011514048.1
XM_017011688.1 Intron XP_016867177.1
XM_017011689.1 Intron XP_016867178.1
XM_017011690.1 Intron XP_016867179.1
XM_017011691.1 Intron XP_016867180.1
XM_017011692.1 Intron XP_016867181.1
XM_017011693.1 Intron XP_016867182.1
XM_017011694.1 Intron XP_016867183.1
XM_017011695.1 Intron XP_016867184.1
XM_017011696.1 Intron XP_016867185.1
XM_017011697.1 Intron XP_016867186.1
XM_017011698.1 Intron XP_016867187.1

View Full Product Details