Product Details
- SNP ID
-
rs16858011
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:183710276 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCATAAAATCTACAAGTCAGAC[A/C]AGAAGAGTCCAGAAGGTAATGTTAT
- Phenotype
-
MIM: 613373
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
YEATS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75748075] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- YEATS2
- Gene Name
- YEATS domain containing 2
View Full Product Details