Product Details

SNP ID
rs12721218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:148701720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAAATGGGTAAAAAATGAGTGT[A/G]CCTTGTTTCCTATTATTACAATGTG
Phenotype
MIM: 106165
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGTR1 PubMed Links

Gene Details

Gene
AGTR1
Gene Name
angiotensin II receptor type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000685.4 Intron NP_000676.1
NM_004835.4 Intron NP_004826.5
NM_009585.3 Intron NP_033611.1
NM_031850.3 Intron NP_114038.4
NM_032049.3 Intron NP_114438.2

View Full Product Details