Product Details

SNP ID
rs35928427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:64581663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCACGCTGCTGCTCGCCAAGAA[A/C]TACATCCTCATGCAGGCGCAGGCCC
Phenotype
MIM: 613483 MIM: 603711
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BHLHE22 PubMed Links

Gene Details

Gene
BHLHE22
Gene Name
basic helix-loop-helix family member e22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152414.4 1426 Missense Mutation AAA,AAC K291N NP_689627.1
Gene
CYP7B1
Gene Name
cytochrome P450 family 7 subfamily B member 1
There are no transcripts associated with this gene.

Gene
LOC401463
Gene Name
uncharacterized LOC401463
There are no transcripts associated with this gene.

View Full Product Details