Product Details

SNP ID
rs17243843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:115146857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTAAGTCATCTGTAAGAATGT[C/T]TACCGTGGAATTCTAGCAAAAGAAA
Phenotype
MIM: 604767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEC1 PubMed Links

Gene Details

Gene
DEC1
Gene Name
deleted in esophageal cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017418.2 Intron NP_059114.1
Gene
LOC101928748
Gene Name
uncharacterized LOC101928748
There are no transcripts associated with this gene.

View Full Product Details