Product Details

SNP ID
rs45504599
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5044528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGCGCGTCCACATCAACATCTC[C/T]GGGCTGCGCTTTGAGACGCAGCTGG
Phenotype
MIM: 176267
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KCNA5 PubMed Links

Gene Details

Gene
KCNA5
Gene Name
potassium voltage-gated channel subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002234.3 610 Silent Mutation TCC,TCT S127S NP_002225.2

View Full Product Details