Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001193465.1 | 2825 | UTR 3 | NP_001180394.1 | ||
NM_001193466.1 | 2825 | UTR 3 | NP_001180395.1 | ||
NM_015443.3 | 2825 | UTR 3 | NP_056258.1 | ||
XM_006721823.1 | 2825 | UTR 3 | XP_006721886.1 | ||
XM_006721824.3 | 2825 | UTR 3 | XP_006721887.1 | ||
XM_011524628.2 | 2825 | UTR 3 | XP_011522930.1 | ||
XM_011524629.2 | 2825 | UTR 3 | XP_011522931.1 | ||
XM_011524630.2 | 2825 | UTR 3 | XP_011522932.1 | ||
XM_011524631.2 | 2825 | UTR 3 | XP_011522933.1 | ||
XM_011524632.2 | 2825 | UTR 3 | XP_011522934.1 | ||
XM_017024488.1 | 2825 | Intron | XP_016879977.1 | ||
XM_017024489.1 | 2825 | Intron | XP_016879978.1 |