Product Details

SNP ID
rs16941050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110848273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGAACAGAACTGGAGGTAGGTTG[A/C]ATTGGCCCCATGGACTATGAGTTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC63 PubMed Links

Gene Details

Gene
CCDC63
Gene Name
coiled-coil domain containing 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286243.1 Intron NP_001273172.1
NM_001286244.1 Intron NP_001273173.1
NM_152591.2 Intron NP_689804.1
XM_006719262.1 Intron XP_006719325.1
XM_006719263.2 Intron XP_006719326.1
XM_011537999.1 Intron XP_011536301.1
XM_011538001.2 Intron XP_011536303.1

View Full Product Details