Product Details

SNP ID
rs35662236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:151695519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGGGCTGCGTGCGCAAACAGGG[C/T]GGCCGCCTTCTGGGCCCGGAGAACG
Phenotype
MIM: 300685
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRRG3 PubMed Links

Gene Details

Gene
PRRG3
Gene Name
proline rich and Gla domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024082.3 64 Intron NP_076987.3
XM_005274747.3 64 UTR 5 XP_005274804.1
XM_011531197.2 64 Intron XP_011529499.1

View Full Product Details