Product Details

SNP ID
rs17031981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154744668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCATCCGCGTGGACACAGTGGA[A/G]GACTTCGCCTACGGAGCTAACATCC
Phenotype
MIM: 604863
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRAT PubMed Links

Gene Details

Gene
LRAT
Gene Name
lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301645.1 401 Silent Mutation GAA,GAG E114E NP_001288574.1
NM_004744.4 401 Silent Mutation GAA,GAG E114E NP_004735.2

View Full Product Details