Product Details

SNP ID
rs6425949
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:35583808 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCACCACTGCCCTTCTGTGACC[C/G]ACCTGCTTCCCTAACCATGGATTCC
Phenotype
MIM: 614428
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TFAP2E PubMed Links

Gene Details

Gene
TFAP2E
Gene Name
transcription factor AP-2 epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178548.3 Intron NP_848643.2
XM_017001139.1 Intron XP_016856628.1

View Full Product Details