Product Details

SNP ID
rs17296821
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46291778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGAATCAGCTTCTTTCTGAGCTG[A/C]CAGAAGTACAGAAGTTGGAGCCTCC
Phenotype
MIM: 603294
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
C21orf58 PubMed Links
Additional Information
For this assay, SNP(s) [rs16979054] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C21orf58
Gene Name
chromosome 21 open reading frame 58
There are no transcripts associated with this gene.

Gene
MCM3AP
Gene Name
minichromosome maintenance complex component 3 associated protein
There are no transcripts associated with this gene.

Gene
YBEY
Gene Name
ybeY metallopeptidase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006114.2 785 UTR 3 NP_001006114.1
NM_001314022.1 785 Intron NP_001300951.1
NM_001314023.1 785 Intron NP_001300952.1
NM_001314024.1 785 Intron NP_001300953.1
NM_001314025.1 785 Intron NP_001300954.1
NM_001314026.1 785 UTR 3 NP_001300955.1
NM_058181.2 785 Intron NP_478061.1
XM_005261156.3 785 Intron XP_005261213.2
XM_011529629.2 785 Intron XP_011527931.1
XM_011529630.2 785 Intron XP_011527932.1
XM_011529631.2 785 Intron XP_011527933.1
XM_011529633.2 785 Intron XP_011527935.1
XM_011529634.2 785 Intron XP_011527936.2
XM_011529635.2 785 Intron XP_011527937.1
XM_017028394.1 785 Intron XP_016883883.1
XM_017028395.1 785 Intron XP_016883884.1
XM_017028396.1 785 Intron XP_016883885.1

View Full Product Details