Product Details

SNP ID
rs17732686
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18274347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCACAGCCATTTCCTTGCATTA[C/T]ACCGTCCTTCTCAGGACGTCTATGA
Phenotype
MIM: 615498 MIM: 601243
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MIEF2 PubMed Links

Gene Details

Gene
MIEF2
Gene Name
mitochondrial elongation factor 2
There are no transcripts associated with this gene.

Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320759.1 3775 UTR 3 NP_001307688.1
NM_004618.4 3775 UTR 3 NP_004609.1
XM_011524001.2 3775 UTR 3 XP_011522303.1

View Full Product Details