Product Details

SNP ID
rs16872562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:42434683 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCATGTTGTTTATCTCTGCCATT[C/T]TTATAGAAGGATGATAGTTAGCTAT
Phenotype
MIM: 600946
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GHR PubMed Links

Gene Details

Gene
GHR
Gene Name
growth hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000163.4 Intron NP_000154.1
NM_001242399.2 Intron NP_001229328.1
NM_001242400.2 Intron NP_001229329.1
NM_001242401.3 Intron NP_001229330.1
NM_001242402.2 Intron NP_001229331.1
NM_001242403.2 Intron NP_001229332.1
NM_001242404.2 Intron NP_001229333.1
NM_001242405.2 Intron NP_001229334.1
NM_001242406.2 Intron NP_001229335.1
NM_001242460.1 Intron NP_001229389.1
NM_001242462.1 Intron NP_001229391.1

View Full Product Details