Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001267895.1 | 1319 | Intron | NP_001254824.1 | ||
NM_133455.3 | 1319 | Intron | NP_597712.2 | ||
XM_005261328.2 | 1319 | Intron | XP_005261385.1 | ||
XM_005261329.2 | 1319 | Intron | XP_005261386.1 | ||
XM_011529868.2 | 1319 | Intron | XP_011528170.1 | ||
XM_011529869.2 | 1319 | Intron | XP_011528171.1 | ||
XM_011529870.2 | 1319 | Intron | XP_011528172.1 | ||
XM_011529871.2 | 1319 | Intron | XP_011528173.1 | ||
XM_011529872.2 | 1319 | Intron | XP_011528174.1 | ||
XM_011529873.2 | 1319 | Intron | XP_011528175.1 | ||
XM_011529874.2 | 1319 | Intron | XP_011528176.1 | ||
XM_011529875.2 | 1319 | Intron | XP_011528177.1 | ||
XM_011529876.1 | 1319 | Intron | XP_011528178.1 | ||
XM_017028589.1 | 1319 | UTR 5 | XP_016884078.1 |