Product Details

SNP ID
rs17849991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29767425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGGCCGCGACGTTGACTTCGAA[A/G]TTGTACTCCCTGCTGTTCCGCAGGA
Phenotype
MIM: 610843
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UQCR10 PubMed Links

Gene Details

Gene
UQCR10
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003684.1 57 Silent Mutation AAA,AAG K9K NP_001003684.1
NM_013387.3 57 Silent Mutation AAA,AAG K9K NP_037519.2
Gene
ZMAT5
Gene Name
zinc finger matrin-type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003692.1 57 Intron NP_001003692.1
NM_001318129.1 57 Intron NP_001305058.1
NM_019103.2 57 Intron NP_061976.1

View Full Product Details