Product Details

SNP ID
rs17001869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40382969 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACACTGTATAAAGGTGAAGGTAGA[C/T]TTCTTCAGAAAGTCCTGAAATACTA
Phenotype
MIM: 608222 MIM: 610440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADSL PubMed Links

Gene Details

Gene
ADSL
Gene Name
adenylosuccinate lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000026.3 Intron NP_000017.1
NM_001123378.2 Intron NP_001116850.1
NM_001317923.1 Intron NP_001304852.1
XM_011529977.2 Intron XP_011528279.1
XM_011529979.1 Intron XP_011528281.1
XM_011529980.2 Intron XP_011528282.1
XM_017028636.1 Intron XP_016884125.1
XM_017028637.1 Intron XP_016884126.1
XM_017028638.1 Intron XP_016884127.1
XM_017028639.1 Intron XP_016884128.1
XM_017028640.1 Intron XP_016884129.1
Gene
SGSM3
Gene Name
small G protein signaling modulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301849.1 Intron NP_001288778.1
NM_015705.5 Intron NP_056520.2
XM_005261572.2 Intron XP_005261629.1
XM_005261573.3 Intron XP_005261630.1
XM_005261574.4 Intron XP_005261631.1
XM_005261575.1 Intron XP_005261632.1
XM_011530158.1 Intron XP_011528460.1
XM_011530159.1 Intron XP_011528461.1
XM_011530160.1 Intron XP_011528462.1
XM_011530161.1 Intron XP_011528463.1
XM_011530162.2 Intron XP_011528464.1
XM_017028775.1 Intron XP_016884264.1
XM_017028776.1 Intron XP_016884265.1
XM_017028777.1 Intron XP_016884266.1
XM_017028778.1 Intron XP_016884267.1
XM_017028779.1 Intron XP_016884268.1
XM_017028780.1 Intron XP_016884269.1

View Full Product Details