Product Details

SNP ID
rs17804214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151826963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATTACAAAAAGCAGCTAGTGT[A/G]TAGGGAAAATGGAAAAATAAAATCT
Phenotype
MIM: 138491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLRA1 PubMed Links

Gene Details

Gene
GLRA1
Gene Name
glycine receptor alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000171.3 Intron NP_000162.2
NM_001146040.1 Intron NP_001139512.1
NM_001292000.1 Intron NP_001278929.1
XM_017009349.1 Intron XP_016864838.1

View Full Product Details