Product Details

SNP ID
rs16909158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93457882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCTGCCTCCTTGAACCTTTTTC[A/G]ATGATCAAAATGCTTCCATTTCCTG
Phenotype
MIM: 612265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM120A PubMed Links

Gene Details

Gene
FAM120A
Gene Name
family with sequence similarity 120A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286722.1 Intron NP_001273651.1
NM_001286723.1 Intron NP_001273652.1
NM_001286724.1 Intron NP_001273653.1
NM_014612.4 Intron NP_055427.2
XM_005251842.4 Intron XP_005251899.1
XM_011518412.2 Intron XP_011516714.1
XM_011518413.2 Intron XP_011516715.1
XM_011518414.2 Intron XP_011516716.1
XM_011518416.2 Intron XP_011516718.1
Gene
FAM120AOS
Gene Name
family with sequence similarity 120A opposite strand
There are no transcripts associated with this gene.

View Full Product Details