Product Details

SNP ID
rs17033616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:35646224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGCCCATAAATTCAATGGAAAA[A/G]TGGAAGACTAACACATTTATTTTTC
Phenotype
MIM: 605488
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPP21 PubMed Links

Gene Details

Gene
ARPP21
Gene Name
cAMP regulated phosphoprotein 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025068.1 Intron NP_001020239.1
NM_001025069.1 Intron NP_001020240.1
NM_001267616.1 Intron NP_001254545.1
NM_001267617.1 Intron NP_001254546.1
NM_001267618.1 Intron NP_001254547.1
NM_001267619.1 Intron NP_001254548.1
NM_016300.4 Intron NP_057384.2
NM_198399.1 Intron NP_938409.1
XM_005264811.2 Intron XP_005264868.1
XM_006712943.2 Intron XP_006713006.1
XM_006712944.2 Intron XP_006713007.1
XM_011533298.2 Intron XP_011531600.1
XM_011533299.2 Intron XP_011531601.1
XM_011533300.2 Intron XP_011531602.1
XM_011533301.2 Intron XP_011531603.1
XM_011533302.2 Intron XP_011531604.1
XM_011533303.2 Intron XP_011531605.1
XM_017005574.1 Intron XP_016861063.1
XM_017005575.1 Intron XP_016861064.1
XM_017005576.1 Intron XP_016861065.1
XM_017005577.1 Intron XP_016861066.1
XM_017005578.1 Intron XP_016861067.1
XM_017005579.1 Intron XP_016861068.1
XM_017005580.1 Intron XP_016861069.1
XM_017005581.1 Intron XP_016861070.1
XM_017005582.1 Intron XP_016861071.1
XM_017005583.1 Intron XP_016861072.1
XM_017005584.1 Intron XP_016861073.1
XM_017005585.1 Intron XP_016861074.1
XM_017005586.1 Intron XP_016861075.1
XM_017005587.1 Intron XP_016861076.1
XM_017005588.1 Intron XP_016861077.1
XM_017005589.1 Intron XP_016861078.1
XM_017005590.1 Intron XP_016861079.1
XM_017005591.1 Intron XP_016861080.1
XM_017005592.1 Intron XP_016861081.1
XM_017005593.1 Intron XP_016861082.1
XM_017005594.1 Intron XP_016861083.1
XM_017005595.1 Intron XP_016861084.1
XM_017005596.1 Intron XP_016861085.1
XM_017005597.1 Intron XP_016861086.1
XM_017005598.1 Intron XP_016861087.1
XM_017005599.1 Intron XP_016861088.1
XM_017005600.1 Intron XP_016861089.1
XM_017005601.1 Intron XP_016861090.1
XM_017005602.1 Intron XP_016861091.1
XM_017005603.1 Intron XP_016861092.1
XM_017005604.1 Intron XP_016861093.1
XM_017005605.1 Intron XP_016861094.1
XM_017005606.1 Intron XP_016861095.1
XM_017005607.1 Intron XP_016861096.1
XM_017005608.1 Intron XP_016861097.1
XM_017005609.1 Intron XP_016861098.1
XM_017005610.1 Intron XP_016861099.1
XM_017005611.1 Intron XP_016861100.1
XM_017005612.1 Intron XP_016861101.1

View Full Product Details