Product Details
- SNP ID
-
rs17851131
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:22987919 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAACCTACCCGATATCGGCGACTT[A/C]GAAGTTTCTTCTCCTCTTTTTCCTT
- Phenotype
-
MIM: 609066
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AJUBA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116754674] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AJUBA
- Gene Name
- ajuba LIM protein
There are no transcripts associated with this gene.
- Gene
- C14orf93
- Gene Name
- chromosome 14 open reading frame 93
View Full Product Details