Product Details

SNP ID
rs17125731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23025025 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTCTGTGAATGACTTCTCCAAA[C/T]CTCTCAGTCTCAAAACATGACACTG
Phenotype
MIM: 600306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB5 PubMed Links

Gene Details

Gene
PSMB5
Gene Name
proteasome subunit beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130725.1 Intron NP_001124197.1
NM_001144932.2 Intron NP_001138404.1
NM_002797.4 Intron NP_002788.1
XM_005267871.2 Intron XP_005267928.1

View Full Product Details