Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145399.1 | 3199 | UTR 3 | NP_001138871.1 | ||
NM_001584.2 | 3199 | Intron | NP_001575.1 | ||
XM_005253110.3 | 3199 | Intron | XP_005253167.1 | ||
XM_005253111.2 | 3199 | Intron | XP_005253168.1 | ||
XM_005253112.1 | 3199 | Intron | XP_005253169.1 | ||
XM_005253114.3 | 3199 | Intron | XP_005253171.1 | ||
XM_011520346.1 | 3199 | Intron | XP_011518648.1 | ||
XM_011520347.2 | 3199 | Intron | XP_011518649.1 | ||
XM_017018231.1 | 3199 | Intron | XP_016873720.1 | ||
XM_017018232.1 | 3199 | UTR 3 | XP_016873721.1 | ||
XM_017018233.1 | 3199 | Intron | XP_016873722.1 | ||
XM_017018234.1 | 3199 | Intron | XP_016873723.1 | ||
XM_017018235.1 | 3199 | Intron | XP_016873724.1 | ||
XM_017018236.1 | 3199 | Intron | XP_016873725.1 | ||
XM_017018237.1 | 3199 | Intron | XP_016873726.1 |