Product Details

SNP ID
rs16858606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218060849 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCTCATAGCAGACTGGGCTAACA[C/T]TGGATGGGTAGAAGGTCCTTCGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCR2P1 PubMed Links

Gene Details

Gene
CXCR2P1
Gene Name
C-X-C motif chemokine receptor 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
RUFY4
Gene Name
RUN and FYVE domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198483.3 Intron NP_940885.2
XM_011511014.2 Intron XP_011509316.1
XM_017003895.1 Intron XP_016859384.1
XM_017003896.1 Intron XP_016859385.1
XM_017003897.1 Intron XP_016859386.1

View Full Product Details