Product Details

SNP ID
rs17850893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76303073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAAGGATGGCGTGGTGGAGATC[A/T]CCGGTGAGCCCCCCTGCTCCTGCAG
Phenotype
MIM: 602195
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HSPB1 PubMed Links

Gene Details

Gene
HSPB1
Gene Name
heat shock protein family B (small) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001540.3 516 Missense Mutation ACC,TCC T121S NP_001531.1

View Full Product Details